![Difloxacin hydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/144/706/8351f129-d97f-4296-8dfb-ad3af9ce6054/640/8351f129-d97f-4296-8dfb-ad3af9ce6054.png)
Difloxacin hydrochloride
![Difloxacin hydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/144/706/8351f129-d97f-4296-8dfb-ad3af9ce6054/640/8351f129-d97f-4296-8dfb-ad3af9ce6054.png)
Synonym(s):
6-Fluoro-1-(4-fluorophenyl)-1,4-dihydro-7-(4-methylpiperazino)-4-oxo-3-quinolinecarboxylic acid hydrochloride
Empirical Formula (Hill Notation):
C21H19F2N3O3 · HCl
CAS Number:
Molecular Weight:
435.85
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing