Skip to Content
Merck
Diethyl malonate ReagentPlus®, 99%

Diethyl malonate

Diethyl malonate ReagentPlus®, 99%
Synonym(s):
Malonic acid diethyl ester, Diethyl malonate
Linear Formula:
CH2(COOC2H5)2
CAS Number:
Molecular Weight:
160.17

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing