Skip to Content
Merck
Dibutyl itaconate analytical standard

Dibutyl itaconate

Dibutyl itaconate analytical standard
Synonym(s):
2-Methylenebutanedioic acid1,4-dibutyl ester
Linear Formula:
CH3(CH2)3O2CCH2C(=CH2)CO2(CH2)3CH3
CAS Number:
Molecular Weight:
242.31

Products

Brand
Boiling Point (°C)
Formula Weight
Purity
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing