Dibromochloromethane
Synonym(s):
Chlorodibromomethane, Dibromochloromethane
Linear Formula:
Br2CHCl
CAS Number:
Molecular Weight:
208.28
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Package Size
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing