Skip to Content
Merck
Diazinon-(diethyl-d10) ≥99 atom % D, ≥98% (CP)

Diazinon-(diethyl-d10)

Diazinon-(diethyl-d10) ≥99 atom % D, ≥98% (CP)
Empirical Formula (Hill Notation):
C12D10H11N2O3PS
CAS Number:
Molecular Weight:
314.41

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Product Line
Purity
Format
Quality Segment
Isotope
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2