Skip to Content
Merck
dGTP =98%, solution, 100 mM, suitable for PCR, suitable for DNA sequencing

dGTP

dGTP =98%, solution, 100 mM, suitable for PCR, suitable for DNA sequencing
Synonym(s):
2′-Deoxyguanosine 5′-triphosphate trisodium salt solution, dGTP-Na3
Empirical Formula (Hill Notation):
C10H13N5Na3O13P3
CAS Number:
Molecular Weight:
573.13

Products

Formula Weight
Manufacturer
Physical Form
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing