
Dehydroandrographolide

Synonym(s):
(1R-(1a,2a,4aa,5β(E),8aβ))-3-[2-(Decahydro-6-hydroxy-5-(hydroxymethyl)-5,8a-dimethyl-2-methylene-1-naphthalenyl)ethylidene)-2(3H)-furanone
Empirical Formula (Hill Notation):
C20H28O4
CAS Number:
Molecular Weight:
332.43
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing