![Deferoxamine Mesylate certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/291/628/3ae6a617-60d4-4658-bfd3-1ede2c0da249/640/3ae6a617-60d4-4658-bfd3-1ede2c0da249.png)
Deferoxamine mesylate
![Deferoxamine Mesylate certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/291/628/3ae6a617-60d4-4658-bfd3-1ede2c0da249/640/3ae6a617-60d4-4658-bfd3-1ede2c0da249.png)
Synonym(s):
Deferoxamine mesylate salt, DFOM, Deferoxamine methanesulfonate salt, Desferrioxamine mesylate salt
Empirical Formula (Hill Notation):
C25H48N6O8 · CH4O3S
CAS Number:
Molecular Weight:
656.79
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-2 of 2