
(+)-Dihydroquinidine

Synonym(s):
Hydroquinidine, (9S)-10,11-Dihydro-6′-methoxycinchonan-9-ol, Dihydroquinidine
Empirical Formula (Hill Notation):
C20H26N2O2
CAS Number:
Molecular Weight:
326.43
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing