Skip to Content
Merck
DBP 98% (HPLC)

DBP

DBP 98% (HPLC)
Synonym(s):
5,10,15,20-Tetraphenylbisbenz[5,6]indeno[1,2,3-cd:1′,2′,3′-lm]perylene, Dibenzo{[f,f′]-4,4′,7,7′-tetraphenyl}diindeno[1,2,3-cd:1′,2′,3′-lm]perylene, Red 2
Empirical Formula (Hill Notation):
C64H36
CAS Number:
Molecular Weight:
804.97

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing