Skip to Content
Merck
(+)-Biotin N-hydroxysuccinimide ester ≥98% (HPLC), powder

(+)-Biotin N-hydroxysuccinimide ester

(+)-Biotin N-hydroxysuccinimide ester ≥98% (HPLC), powder
Synonym(s):
(+)-Biotin N-succinimidyl ester, d-Biotin NHS ester, BNHS, Biotinyl-N-hydroxy-succinimide, N-Hydroxysuccinimidobiotin, N-Succinimidyl D-biotinate, NHS-Biotin
Empirical Formula (Hill Notation):
C14H19N3O5S
CAS Number:
Molecular Weight:
341.38

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing