Skip to Content
Merck
Corning® Transwell® polyester membrane cell culture inserts 6.5 mm Transwell with 3.0 μm pore polyester membrane insert, TC-treated, sterile, 48/cs

Corning® Transwell® 24 well plates

Corning® Transwell® polyester membrane cell culture inserts 6.5 mm Transwell with 3.0 μm pore polyester membrane insert, TC-treated, sterile, 48/cs

Products

Product Category
Color
Manufacturer
Binding Type
Diameter (mm)
Surface Area cm²
Wells
Working Volume (ml)
Color
Membrane Material
Plate Material
Package Size
Pore size (µm)
Sterility
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing