![Clomiphene citrate United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/270/453/9a845282-f10e-478b-b2ff-6d87abc1cb21/640/9a845282-f10e-478b-b2ff-6d87abc1cb21.png)
Clomiphene citrate
![Clomiphene citrate United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/270/453/9a845282-f10e-478b-b2ff-6d87abc1cb21/640/9a845282-f10e-478b-b2ff-6d87abc1cb21.png)
Synonym(s):
Clomiphene citrate salt, 2-(4-[2-Chloro-1,2-diphenylethenyl]phenoxy)-N,N-diethylethanamine
Empirical Formula (Hill Notation):
C26H28ClNO · C6H8O7
CAS Number:
Molecular Weight:
598.08
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing