![Clofilium tosylate >97%, solid](/deepweb/assets/sigmaaldrich/product/structures/235/066/82319718-e9d9-4fc6-85ee-6b13149a6f1b/640/82319718-e9d9-4fc6-85ee-6b13149a6f1b.png)
Clofilium tosylate
![Clofilium tosylate >97%, solid](/deepweb/assets/sigmaaldrich/product/structures/235/066/82319718-e9d9-4fc6-85ee-6b13149a6f1b/640/82319718-e9d9-4fc6-85ee-6b13149a6f1b.png)
Synonym(s):
4-Chloro-N,N-diethyl-N-heptylbenzenebutanaminium tosylate
Empirical Formula (Hill Notation):
C21H37ClN · C7H7O3S
CAS Number:
Molecular Weight:
510.17
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing