Skip to Content
CH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Get Site Smart
Back
Chromolith
®
WP 300 Epoxy (monolithic) HPLC Columns
Products
Brand
Supelco (7)
pH Value
Product Line
ID (mm)
Length (cm)
Quality Segment
Column Material
Package Size
Pore size (µm)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
1.52251
Chromolith
®
WP 300 Epoxy (monolithic) HPLC Columns,
L × I.D. 50 mm x 4.6 mm HPLC Column
Expand
View Pricing
1.52250
Chromolith
®
WP 300 Epoxy (monolithic) HPLC Columns,
L × I.D. 100 mm x 4.6 mm HPLC Column
Expand
View Pricing
1.52252
Chromolith
®
WP 300 Epoxy (monolithic) HPLC Columns,
L × I.D. 25 mm x 4.6 mm HPLC Column
Expand
View Pricing
1.52352
Chromolith
®
WP 300 Epoxy (monolithic) HPLC Columns,
L × I.D. 25 mm × 2 mm HPLC Column
Expand
View Pricing
1.52350
Chromolith
®
WP 300 Epoxy (monolithic) HPLC Columns,
L × I.D. 10 cm × 2 mm HPLC Column
Expand
View Pricing
1.52253
Chromolith
®
WP 300 Epoxy (monolithic) HPLC Columns,
L × I.D. 10 mm x 4.6 mm Guard Cartridges (3 pieces)
Expand
View Pricing
1.52254
Chromolith
®
WP 300 Epoxy (monolithic) HPLC Columns,
L × I.D. 5 mm x 4.6 mm Guard Cartridges (3 pieces)
Expand
View Pricing