Skip to Content
Merck
Cetirizine hydrochloride British Pharmacopoeia (BP) Reference Standard

Cetirizine Hydrochloride

Cetirizine hydrochloride British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Cetirizine dihydrochloride, [2-[4-[(4-Chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]acetic acid dihydrochloride
Empirical Formula (Hill Notation):
C21H25ClN2O3 · 2HCl
CAS Number:
Molecular Weight:
461.81

Products

Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing