Skip to Content
Merck
vibrant-m

Cefaclor

vibrant-m
Synonym(s):
Cefaclor monohydrate
Empirical Formula (Hill Notation):
C15H14ClN3O4S · H2O
CAS Number:
Molecular Weight:
385.82
EC Number:

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique