
Carboxy-PTIO potassium salt

Synonym(s):
2-(4-Carboxyphenyl)-4,4,5,5-tetramethylimidazoline-1-oxyl-3-oxide potassium salt, 2-(4-Carboxyphenyl)-4,5-dihydro-4,4,5,5-tetramethyl-1H-imidazol-1-yloxy-3-oxide potassium salt
Empirical Formula (Hill Notation):
C14H16KN2O4
CAS Number:
Molecular Weight:
315.39
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing