![Carboplatin British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/340/300/021af797-1f9b-42c3-a871-5298bfc9f235/640/021af797-1f9b-42c3-a871-5298bfc9f235.png)
Carboplatin
![Carboplatin British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/340/300/021af797-1f9b-42c3-a871-5298bfc9f235/640/021af797-1f9b-42c3-a871-5298bfc9f235.png)
Synonym(s):
cis-Diammine(1,1-cyclobutanedicarboxylato) platinum, Carboplatin
Linear Formula:
[C4H6(CO2)2]Pt(NH3)2
CAS Number:
Molecular Weight:
371.25
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Activity Spectrum
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing