Skip to Content
Merck
Bruker standard solvent suitable for MALDI-MS, 50% ACN + 47.5% H2O + 2.5% TFA

Bruker standard solvent

Bruker standard solvent suitable for MALDI-MS, 50% ACN + 47.5% H2O + 2.5% TFA
Synonym(s):
Trifluoroacetic acid solution, TFA
Linear Formula:
CF3COOH
CAS Number:
Molecular Weight:
114.02

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing