Skip to Content
Merck
Bromobutide certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Bromobutide

Bromobutide certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
(RS)-2-Bromo-3,3-dimethyl-N-(1-methyl-1-phenylethyl)butyramide, 2-Bromo-3,3-dimethyl-N-(α,α-dimethylbenzyl)butyramide, 2-Bromo-3,3-dimethyl-N-(1-methyl-1-phenylethyl)butanamide
Empirical Formula (Hill Notation):
C15H22BrNO
CAS Number:
Molecular Weight:
312.25

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Technique