Skip to Content
Merck
Bromobenzene ReagentPlus®, 99%

Bromobenzene

Bromobenzene ReagentPlus®, 99%
Synonym(s):
Bromobenzene
Empirical Formula (Hill Notation):
C6H5Br
CAS Number:
Molecular Weight:
157.01

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing