![Bretylium tosylate United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/295/290/1393e246-d71a-4eae-baa9-e3326c55c6f9/640/1393e246-d71a-4eae-baa9-e3326c55c6f9.png)
Bretylium tosylate
![Bretylium tosylate United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/295/290/1393e246-d71a-4eae-baa9-e3326c55c6f9/640/1393e246-d71a-4eae-baa9-e3326c55c6f9.png)
Synonym(s):
(o-Bromobenzyl)ethyldimethylammonium p-toluenesulfonate
Empirical Formula (Hill Notation):
C18H24BrNO3S
CAS Number:
Molecular Weight:
414.36
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing