Skip to Content
Merck
BIS-TRIS propane anhydrous, free-flowing, Redi-Dri™, ≥99.0%

BIS-TRIS propane

BIS-TRIS propane anhydrous, free-flowing, Redi-Dri™, ≥99.0%
Synonym(s):
1,3-Bis[tris(hydroxymethyl)methylamino]propane, BIS-TRIS propane, BIS-Tris Propane, ULTROL® Grade
Linear Formula:
CH2[CH2NHC(CH2OH)3]2
CAS Number:
Molecular Weight:
282.33
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Research Area
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique