Skip to Content
Merck
Bis(trimethylsilyl) malonate 98%

Bis(trimethylsilyl) malonate

Bis(trimethylsilyl) malonate 98%
Synonym(s):
Malonic acid bis(trimethylsilyl ester)
Linear Formula:
(CH3)3SiOCOCH2COOSi(CH3)3
CAS Number:
Molecular Weight:
248.42
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing