Skip to Content
Merck
Bispyribac sodium salt PESTANAL®, analytical standard

Bispyribac sodium salt

Bispyribac sodium salt PESTANAL®, analytical standard
Synonym(s):
Sodium 2,6-bis(4,6-dimethoxy-2-pyrimidinyloxy)benzoate
Empirical Formula (Hill Notation):
C19H17N4NaO8
CAS Number:
Molecular Weight:
452.35

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2