Skip to Content
Merck
Betulin analytical standard

Betulin

Betulin analytical standard
Synonym(s):
Betulinic alcohol, Betulinol, Betulol, Lup-20(29)-ene-3β,28-diol, Trochol
Empirical Formula (Hill Notation):
C30H50O2
CAS Number:
Molecular Weight:
442.72
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing