Skip to Content
Merck
Betahistine hydrochloride United States Pharmacopeia (USP) Reference Standard

Betahistine Hydrochloride

Betahistine hydrochloride United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Betahistine dihydrochloride, Methyl[2-(2-pyridyl)ethyl]amine dihydrochloride
Empirical Formula (Hill Notation):
C8H12N2 · 2HCl
CAS Number:
Molecular Weight:
209.12

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Special Grade