![Benzoyl peroxide blend with dicyclohexyl phthalate suitable for use as a catalyst for electron microscopy. Modified to render it safe in transit.](/deepweb/assets/sigmaaldrich/product/structures/282/727/a56757ba-f566-489f-845a-28b8c50da6bc/640/a56757ba-f566-489f-845a-28b8c50da6bc.png)
Benzoyl peroxide blend with dicyclohexyl phthalate
![Benzoyl peroxide blend with dicyclohexyl phthalate suitable for use as a catalyst for electron microscopy. Modified to render it safe in transit.](/deepweb/assets/sigmaaldrich/product/structures/282/727/a56757ba-f566-489f-845a-28b8c50da6bc/640/a56757ba-f566-489f-845a-28b8c50da6bc.png)
Empirical Formula (Hill Notation):
C14H10O4
Molecular Weight:
242.23
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing