Benzo[a]pyrene-d12
Synonym(s):
3,4-Benzopyrene-d12
Empirical Formula (Hill Notation):
C20D12
CAS Number:
Molecular Weight:
264.38
Products
Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing