![Benzalkonium chloride for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/711/355/4044fc12-fb54-4100-b4c3-89ba15b4ad88/640/4044fc12-fb54-4100-b4c3-89ba15b4ad88.png)
Benzalkonium chloride for system suitability
![Benzalkonium chloride for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/711/355/4044fc12-fb54-4100-b4c3-89ba15b4ad88/640/4044fc12-fb54-4100-b4c3-89ba15b4ad88.png)
Synonym(s):
Benzalkonium chloride, Alkylbenzyldimethylammonium chloride, Alkyldimethylbenzylammonium chloride
Linear Formula:
C6H5CH2N(CH3)2RCl (R=C8H17 to C18H37)
CAS Number:
Products
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing