![Benfuracarb PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/298/307/51e12420-5fb0-4f83-a584-953371a88ada/640/51e12420-5fb0-4f83-a584-953371a88ada.png)
Benfuracarb
![Benfuracarb PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/298/307/51e12420-5fb0-4f83-a584-953371a88ada/640/51e12420-5fb0-4f83-a584-953371a88ada.png)
Empirical Formula (Hill Notation):
C20H30N2O5S
CAS Number:
Molecular Weight:
410.53
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2