Skip to Content
Merck
vibrant-m

Bendamustine Related Compound E

vibrant-m
Synonym(s):
4-{5-[(2-Chloroethyl)(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazol-2-yl}butanoic acid
Empirical Formula (Hill Notation):
C16H22ClN3O3
CAS Number:
Molecular Weight:
339.82

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Showing 1-1 of 1