![β-Cyfluthrin certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/143/868/404af91b-681a-487b-a6a1-7a378e17c422/640/404af91b-681a-487b-a6a1-7a378e17c422.png)
β-Cyfluthrin
![β-Cyfluthrin certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/143/868/404af91b-681a-487b-a6a1-7a378e17c422/640/404af91b-681a-487b-a6a1-7a378e17c422.png)
Empirical Formula (Hill Notation):
C22H18Cl2FNO3
CAS Number:
Molecular Weight:
434.29
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2