![Azadirachtin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/270/847/c02d5d67-1a0b-455d-982e-76d03187da46/640/c02d5d67-1a0b-455d-982e-76d03187da46.png)
Azadirachtin
![Azadirachtin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/270/847/c02d5d67-1a0b-455d-982e-76d03187da46/640/c02d5d67-1a0b-455d-982e-76d03187da46.png)
Empirical Formula (Hill Notation):
C35H44O16
CAS Number:
Molecular Weight:
720.71
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing