![Articaine Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/113/175/cc51b532-e052-4e9a-bb14-f771d6d4480f/640/cc51b532-e052-4e9a-bb14-f771d6d4480f.jpg)
Articaine Related Compound B
![Articaine Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/113/175/cc51b532-e052-4e9a-bb14-f771d6d4480f/640/cc51b532-e052-4e9a-bb14-f771d6d4480f.jpg)
Synonym(s):
4-Methyl-3-[2-(propylamino)propionamido]-2-thiophenecarboxylic acid, 2-Carboxyarticaine, Articainic acid
Empirical Formula (Hill Notation):
C12H18N2O3S
CAS Number:
Molecular Weight:
270.35
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2