Anti-Vinculin Antibody
Synonym(s):
Monoclonal Anti-Vinculin antibody produced in mouse, Anti-VCL, Anti-CMD1W, Anti-MVCL, Metavinculin, VCL, VINC, Anti-Metavinculin antibody produced in rabbit, Anti-Vinculin antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Physical Form
Product Line
Recombinant Host
Antibody Form
Research Area
Conjugate
Quality Segment
Application
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-13 of 13
Compare
Product Number
Description
Pricing