
Anti-SEC61B antibody produced in rabbit

Synonym(s):
Anti-Sec61 beta subunit
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing