
Anti-phospho-CD3ζ (pTyr142) antibody produced in rabbit

Synonym(s):
Anti-CD247, Anti-CD3H, Anti-CD3Q, Anti-CD3Z, Anti-T cell receptor T3 zeta chain, Anti-T3Z, Anti-TCRK, Anti-Tcrz
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing