Skip to Content
Merck
Anti-KHDRBS3 antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-KHDRBS3 antibody produced in rabbit

Anti-KHDRBS3 antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-KH domain-containing, RNA-binding, signal transduction-associated protein 3 antibody produced in rabbit, Anti-RNA-binding protein T-Star antibody produced in rabbit, Anti-SLM-2 antibody produced in rabbit, Anti-Sam68-like mammalian protein 2 antibody produced in rabbit, Anti-Sam68-like phosphotyrosine protein antibody produced in rabbit

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique