![Anti-DLAT antibody produced in rabbit IgG fraction of antiserum](/deepweb/assets/sigmaaldrich/product/images/141/717/ef6bcb72-8676-4fbc-a5b0-a93409f02246/640/ef6bcb72-8676-4fbc-a5b0-a93409f02246.jpg)
Anti-DLAT antibody produced in rabbit
![Anti-DLAT antibody produced in rabbit IgG fraction of antiserum](/deepweb/assets/sigmaaldrich/product/images/141/717/ef6bcb72-8676-4fbc-a5b0-a93409f02246/640/ef6bcb72-8676-4fbc-a5b0-a93409f02246.jpg)
Synonym(s):
Anti-DLTA, Anti-Dihydrolipoamide S-acetyltransferase (E2 component of pyruvate dehydrogenase complex), Anti-PDC-E2, Anti-PDCE2
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing