
Anti-AHSG antibody produced in rabbit

Synonym(s):
Anti-α-2-HS-glycoprotein precursor antibody produced in rabbit, Anti-α-2-Z-globulin antibody produced in rabbit, Anti-Ba-α-2-Glycoprotein antibody produced in rabbit, Anti-Fetuin-A antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing