Anti-α-Catenin antibody produced in rabbit
Synonym(s):
Anti-α-Catenin
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3