![Antazoline Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/195/236/f8deb51a-a521-4da6-8d9c-b3f197f91eab/640/f8deb51a-a521-4da6-8d9c-b3f197f91eab.jpg)
Antazoline Related Compound A
![Antazoline Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/195/236/f8deb51a-a521-4da6-8d9c-b3f197f91eab/640/f8deb51a-a521-4da6-8d9c-b3f197f91eab.jpg)
Synonym(s):
N-(2-Aminoethyl)-2-[benzyl(phenyl)amino]acetamide
Empirical Formula (Hill Notation):
C17H21N3O
CAS Number:
Molecular Weight:
283.37
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2