![Antazoline hydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/213/778/480c373b-b59a-4822-948e-6feeac1918c3/640/480c373b-b59a-4822-948e-6feeac1918c3.png)
Antazoline hydrochloride
![Antazoline hydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/213/778/480c373b-b59a-4822-948e-6feeac1918c3/640/480c373b-b59a-4822-948e-6feeac1918c3.png)
Synonym(s):
2-(N-Benzylanilinomethyl)-2-imidazoline hydrochloride
Empirical Formula (Hill Notation):
C17H19N3 · HCl
CAS Number:
Molecular Weight:
301.81
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing