Skip to Content
Products
Cart
0
CH
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Amyl alcohol
Synonym(s):
1-Pentanol,
n
-Amyl alcohol, Pentyl alcohol
Linear Formula:
CH
3
(CH
2
)
4
OH
CAS Number:
71-41-0
Molecular Weight:
88.15
EC Number:
200-752-1
Products
Product Category
aroma chemicals (1)
Brand
Sigma-Aldrich (1)
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Food Grade (1)
Fragrance grade (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
W205605
Amyl alcohol,
≥99%, FG
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
Français - FR
Deutsch - DE
English - EN
Italiano - IT
Learn More