![AMPSO sodium salt](/deepweb/assets/sigmaaldrich/product/structures/296/187/59592553-5852-445f-bef1-5ff61568fbfa/640/59592553-5852-445f-bef1-5ff61568fbfa.png)
AMPSO sodium salt
![AMPSO sodium salt](/deepweb/assets/sigmaaldrich/product/structures/296/187/59592553-5852-445f-bef1-5ff61568fbfa/640/59592553-5852-445f-bef1-5ff61568fbfa.png)
Synonym(s):
3-([1,1-Dimethyl-2-hydroxyethyl]amino)-2-hydroxypropanesulfonic acid
Linear Formula:
C7H16NO5SNa
CAS Number:
Molecular Weight:
249.26
Products
Product Category
Brand
Formula Weight
pH Value
Physical Form
pK Value
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing