Amlodipine impurity A
Synonym(s):
3-Ethyl 5-methyl (4RS)-4-(2-chlorophenyl)-2-[[2-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)ethoxy]methyl]-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate, 3-Ethyl 5-methyl 4-(2-chlorophenyl)-2-{[2-(1,3-dioxoisoindolin-2-yl)ethoxy]methyl}-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate, Phthalimidoamlodipine
Empirical Formula (Hill Notation):
C28H27ClN2O7
CAS Number:
Molecular Weight:
538.98
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing