Skip to Content
Merck
Aminosalicylic acid United States Pharmacopeia (USP) Reference Standard

Aminosalicylic acid

Aminosalicylic acid United States Pharmacopeia (USP) Reference Standard
Synonym(s):
4-Aminosalicylic acid, 4-Amino-2-hydroxybenzoic acid, PAS
Linear Formula:
H2NC6H3-2-(OH)CO2H
CAS Number:
Molecular Weight:
153.14

Products

Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing