![Aldicarb-sulfoxide certified reference material, TraceCERT®, mixture of stereo isomers, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/148/169/dce4c005-4365-4645-992c-f4b12c27f28d/640/dce4c005-4365-4645-992c-f4b12c27f28d.png)
Aldicarb-sulfoxide
![Aldicarb-sulfoxide certified reference material, TraceCERT®, mixture of stereo isomers, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/148/169/dce4c005-4365-4645-992c-f4b12c27f28d/640/dce4c005-4365-4645-992c-f4b12c27f28d.png)
Empirical Formula (Hill Notation):
C7H14N2O3S
CAS Number:
Molecular Weight:
206.26
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2